Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGCAGGGCCTCAGTCAGGAACAC[A/G]CCCCGCAGGGCCTGCAGCAGGGCGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEDD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DEDD2 - death effector domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270614.1 | 976 | Silent Mutation | GGC,GGT | G,G 277 | NP_001257543.1 | |
NM_001270615.1 | 976 | Silent Mutation | GGC,GGT | G,G 272 | NP_001257544.1 | |
NM_133328.3 | 976 | Silent Mutation | GGC,GGT | G,G 277 | NP_579874.1 | |
XM_011526569.1 | 976 | Silent Mutation | GGC,GGT | G,G 317 | XP_011524871.1 | |
XM_011526571.1 | 976 | UTR 3 | XP_011524873.1 | |||
XM_011526572.1 | 976 | UTR 3 | XP_011524874.1 | |||
XM_017026402.1 | 976 | Silent Mutation | GGC,GGT | G,G 317 | XP_016881891.1 | |
XM_017026403.1 | 976 | UTR 3 | XP_016881892.1 |
POU2F2 - POU class 2 homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |