Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTGGAGCAGCCTCCGCTCCCGC[G/T]CAAACTCCTTCATGCGCATCACATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603842 MIM: 610057 | ||||||||||||||||||||
Literature Links: |
NDUFB7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NDUFB7 - NADH:ubiquinone oxidoreductase subunit B7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004146.5 | 385 | Missense Mutation | GAG,GCG | E,A 103 | NP_004137.2 | |
XM_011528039.2 | 385 | Missense Mutation | GAG,GCG | E,A 65 | XP_011526341.1 |
TECR - trans-2,3-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321170.1 | 385 | Intron | NP_001308099.1 | |||
NM_138501.5 | 385 | Intron | NP_612510.1 | |||
XM_006722945.1 | 385 | Intron | XP_006723008.1 | |||
XM_011528442.2 | 385 | Intron | XP_011526744.1 |