Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGACGTGCCCCCTGCTTCCCGAC[A/G]GTGACCAGCCTCCAGGACCTGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612685 | ||||||||||||||||||||
Literature Links: |
CAMSAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CAMSAP3 - calmodulin regulated spectrin associated protein family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080429.2 | 830 | Silent Mutation | ACA,ACG | T,T 252 | NP_001073898.1 | |
NM_020902.1 | 830 | Silent Mutation | ACA,ACG | T,T 225 | NP_065953.1 | |
XM_005272491.3 | 830 | Silent Mutation | ACA,ACG | T,T 241 | XP_005272548.1 | |
XM_011528154.2 | 830 | Silent Mutation | ACA,ACG | T,T 252 | XP_011526456.1 | |
XM_017027036.1 | 830 | Silent Mutation | ACA,ACG | T,T 252 | XP_016882525.1 |
MIR6792 - microRNA 6792 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |