Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAAAGGTTTTTCTACATACCTTA[C/T]ATTCATAGGGTTTCATACCAGTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF573 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF573 - zinc finger protein 573 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172689.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 440 | NP_001166160.1 | |
NM_001172690.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 528 | NP_001166161.1 | |
NM_001172691.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 526 | NP_001166162.1 | |
NM_001172692.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 440 | NP_001166163.1 | |
NM_152360.3 | 1519 | Missense Mutation | TAT,TGT | Y,C 470 | NP_689573.3 | |
XM_011526446.2 | 1519 | Missense Mutation | TAT,TGT | Y,C 543 | XP_011524748.1 | |
XM_017026275.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 543 | XP_016881764.1 | |
XM_017026276.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 543 | XP_016881765.1 | |
XM_017026277.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 541 | XP_016881766.1 | |
XM_017026278.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 472 | XP_016881767.1 | |
XM_017026279.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 472 | XP_016881768.1 | |
XM_017026280.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 440 | XP_016881769.1 | |
XM_017026281.1 | 1519 | Missense Mutation | TAT,TGT | Y,C 440 | XP_016881770.1 |