Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACTGCACACTGAGTATCCACCC[A/G]GTGCACCTCAATGACAGTGGTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD22 - CD22 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185099.1 | 408 | Silent Mutation | CCA,CCG | P,P 108 | NP_001172028.1 | |
NM_001185100.1 | 408 | Silent Mutation | CCA,CCG | P,P 108 | NP_001172029.1 | |
NM_001185101.1 | 408 | Silent Mutation | CCA,CCG | P,P 108 | NP_001172030.1 | |
NM_001278417.1 | 408 | UTR 5 | NP_001265346.1 | |||
NM_001771.3 | 408 | Silent Mutation | CCA,CCG | P,P 108 | NP_001762.2 |