Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCGGAGTCCACGGTCAGAGCCCC[C/G]TTCCCAGGAATCAGGGGGCACTGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165090 | ||||||||||||||||||||
Literature Links: |
RRAS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRAS - related RAS viral (r-ras) oncogene homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAF1 - SR-related CTD associated factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021228.2 | 46 | Missense Mutation | CCT,CGT | P,R 76 | NP_067051.2 | |
XM_005259122.4 | 46 | Missense Mutation | CCT,CGT | P,R 76 | XP_005259179.1 | |
XM_011527194.2 | 46 | Missense Mutation | CCT,CGT | P,R 79 | XP_011525496.1 | |
XM_017027083.1 | 46 | UTR 5 | XP_016882572.1 |