Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGTCTCAGGCTCCAGGGATAGCGT[A/G]TAGGAGTGATTTTCCTCGGAGCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607320 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM98C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM98C - family with sequence similarity 98 member C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RASGRP4 - RAS guanyl releasing protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146202.1 | 1987 | Silent Mutation | TAC,TAT | Y,Y 611 | NP_001139674.1 | |
NM_001146203.1 | 1987 | Silent Mutation | TAC,TAT | Y,Y 533 | NP_001139675.1 | |
NM_001146204.1 | 1987 | Silent Mutation | TAC,TAT | Y,Y 556 | NP_001139676.1 | |
NM_001146205.1 | 1987 | Silent Mutation | TAC,TAT | Y,Y 591 | NP_001139677.1 | |
NM_001146206.1 | 1987 | Silent Mutation | TAC,TAT | Y,Y 436 | NP_001139678.1 | |
NM_001146207.1 | 1987 | Silent Mutation | TAC,TAT | Y,Y 528 | NP_001139679.1 | |
NM_170604.2 | 1987 | Silent Mutation | TAC,TAT | Y,Y 625 | NP_733749.1 |