Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCTGCAGAGGCTGGAGGCTTATG[G/T]ACGACTCCGTGGTCGCCTCCCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 211100 MIM: 609278 MIM: 609623 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FUT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FUT1 - fucosyltransferase 1 (H blood group) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IZUMO1 - izumo sperm-egg fusion 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321864.1 | 940 | Missense Mutation | NP_001308793.1 | |||
NM_001321865.1 | 940 | Missense Mutation | NP_001308794.1 | |||
NM_182575.2 | 940 | Missense Mutation | NP_872381.2 | |||
XM_005258793.4 | 940 | Missense Mutation | XP_005258850.1 | |||
XM_011526811.2 | 940 | Missense Mutation | XP_011525113.1 | |||
XM_011526812.2 | 940 | Missense Mutation | XP_011525114.1 | |||
XM_011526813.2 | 940 | Missense Mutation | XP_011525115.1 | |||
XM_011526815.2 | 940 | Missense Mutation | XP_011525117.1 | |||
XM_017026646.1 | 940 | Missense Mutation | XP_016882135.1 |
RASIP1 - Ras interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017805.2 | 940 | Intron | NP_060275.2 | |||
XM_011527053.2 | 940 | Intron | XP_011525355.2 | |||
XM_011527054.2 | 940 | Intron | XP_011525356.2 | |||
XM_011527055.2 | 940 | Intron | XP_011525357.2 | |||
XM_017026914.1 | 940 | Intron | XP_016882403.1 | |||
XM_017026915.1 | 940 | Intron | XP_016882404.1 |