Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCTTACTCCTGCGTGAAGGGCA[C/T]AGCAGAAGCAGAGAGGAAGCGCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614545 MIM: 160793 | ||||||||||||||||||||
Literature Links: |
EMC10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EMC10 - ER membrane protein complex subunit 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM71E1 - family with sequence similarity 71 member E1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308429.1 | 691 | Missense Mutation | ATG,GTG | M,V 242 | NP_001295358.1 | |
NM_138411.2 | 691 | Missense Mutation | ATG,GTG | M,V 226 | NP_612420.1 | |
XM_011526405.2 | 691 | Missense Mutation | ATG,GTG | M,V 242 | XP_011524707.2 | |
XM_011526406.2 | 691 | Missense Mutation | ATG,GTG | M,V 226 | XP_011524708.2 |
MYBPC2 - myosin binding protein C, fast type | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |