Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGGGTCTTTTCCACTAAACGGGT[A/C]CCCAAACCCCTTTTTACTTTGGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616826 | ||||||||||||||||||||
Literature Links: |
EPS15L1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPS15L1 - epidermal growth factor receptor pathway substrate 15 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258374.1 | 2564 | Missense Mutation | GAC,TAC | D,Y 833 | NP_001245303.1 | |
NM_001258375.1 | 2564 | UTR 3 | NP_001245304.1 | |||
NM_001258376.1 | 2564 | Intron | NP_001245305.1 | |||
NM_021235.2 | 2564 | Missense Mutation | GAC,TAC | D,Y 833 | NP_067058.1 | |
XM_017027086.1 | 2564 | Missense Mutation | GAC,TAC | D,Y 849 | XP_016882575.1 | |
XM_017027087.1 | 2564 | Missense Mutation | GAC,TAC | D,Y 849 | XP_016882576.1 | |
XM_017027088.1 | 2564 | Intron | XP_016882577.1 | |||
XM_017027089.1 | 2564 | Intron | XP_016882578.1 | |||
XM_017027090.1 | 2564 | Intron | XP_016882579.1 | |||
XM_017027091.1 | 2564 | Intron | XP_016882580.1 | |||
XM_017027092.1 | 2564 | Intron | XP_016882581.1 |