Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGCTGTACCAGCGCTGGGAAC[C/G]GTACCGCAGGGAGTGCCAGGAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137241 | ||||||||||||||||||||
Literature Links: |
GIPR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GIPR - gastric inhibitory polypeptide receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000164.3 | 278 | Missense Mutation | CCG,CGG | P,R 41 | NP_000155.1 | |
NM_001308418.1 | 278 | Missense Mutation | CCG,CGG | P,R 41 | NP_001295347.1 | |
XM_011526709.2 | 278 | Missense Mutation | CCG,CGG | P,R 41 | XP_011525011.1 | |
XM_011526710.2 | 278 | Missense Mutation | CCG,CGG | P,R 41 | XP_011525012.1 | |
XM_011526711.2 | 278 | Missense Mutation | CCG,CGG | P,R 41 | XP_011525013.1 | |
XM_011526713.2 | 278 | Missense Mutation | CCG,CGG | P,R 41 | XP_011525015.1 | |
XM_011526714.2 | 278 | Intron | XP_011525016.1 | |||
XM_011526715.2 | 278 | Intron | XP_011525017.1 | |||
XM_011526716.2 | 278 | Missense Mutation | CCG,CGG | P,R 41 | XP_011525018.1 | |
XM_017026584.1 | 278 | Intron | XP_016882073.1 | |||
XM_017026585.1 | 278 | Intron | XP_016882074.1 | |||
XM_017026586.1 | 278 | Intron | XP_016882075.1 | |||
XM_017026587.1 | 278 | Intron | XP_016882076.1 |
MIR642A - microRNA 642a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR642B - microRNA 642b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |