Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGAAGAATGAAGAAATTCAGAG[C/G]TTTATCTACTCCCTGAAACAGATGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609828 MIM: 614715 | ||||||||||||||||||||
Literature Links: |
FSD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FSD1 - fibronectin type III and SPRY domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024333.2 | 224 | Missense Mutation | AGC,AGG | S,R 24 | NP_077309.1 | |
XM_005259644.2 | 224 | Missense Mutation | AGC,AGG | S,R 24 | XP_005259701.1 |
TMIGD2 - transmembrane and immunoglobulin domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |