Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCATGCTCCGGCTAGGGGTCCGG[A/G]TGGGCTTGTCGGAAGATCGGAAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601768 | ||||||||||||||||||||
Literature Links: |
MPND PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MPND - MPN domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3GL1 - SH3 domain containing GRB2 like 1, endophilin A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199943.1 | Intron | NP_001186872.1 | ||||
NM_001199944.1 | Intron | NP_001186873.1 | ||||
NM_003025.3 | Intron | NP_003016.1 | ||||
XM_011528183.2 | Intron | XP_011526485.1 | ||||
XM_017027146.1 | Intron | XP_016882635.1 |