Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAACTTTGGGTGGCTTGGCTTCA[A/G]TTATTACTTGGGCAGTGGTGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176395 MIM: 176396 | ||||||||||||||||||||
Literature Links: |
PSG6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSG6 - pregnancy specific beta-1-glycoprotein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031850.3 | 245 | Missense Mutation | ACT,ATT | T,I 38 | NP_001027020.1 | |
NM_002782.4 | 245 | Intron | NP_002773.1 |
PSG7 - pregnancy specific beta-1-glycoprotein 7 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |