Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCCAAGGCTCAGTTGGACCCAT[C/G]ATTGCAGGGCTTGGTGATGTTTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100506634 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100506634 - hepatocellular carcinoma-associated antigen HCA25a | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF584 - zinc finger protein 584 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318002.1 | 509 | Intron | NP_001304931.1 | |||
NM_173548.2 | 509 | Missense Mutation | TCA,TGA | S,* 12 | NP_775819.1 | |
XM_005258628.4 | 509 | Intron | XP_005258685.1 | |||
XM_005258630.4 | 509 | Nonsense Mutation | TCA,TGA | S,* 12 | XP_005258687.1 |