Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTTCCCTGCAGAAATTCTGAGC[A/G]GAGTTATTCGGAGTGTCAAGAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614770 MIM: 601717 MIM: 610850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PCP2 - Purkinje cell protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PET100 - PET100 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STXBP2 - syntaxin binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127396.2 | 98 | Missense Mutation | AGA,GGA | R,G 17 | NP_001120868.1 | |
NM_001272034.1 | 98 | Missense Mutation | AGA,GGA | R,G 17 | NP_001258963.1 | |
NM_006949.3 | 98 | Missense Mutation | AGA,GGA | R,G 17 | NP_008880.2 | |
XM_011528210.1 | 98 | Missense Mutation | AGA,GGA | R,G 17 | XP_011526512.1 | |
XM_011528212.2 | 98 | Missense Mutation | AGA,GGA | R,G 17 | XP_011526514.1 |
XAB2 - XPA binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |