Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTCGCCTTCCCCGCCTTACTT[C/T]CATGTCTGTACTGGGCACCTCCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602717 MIM: 131235 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRIN2D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRIN2D - glutamate ionotropic receptor NMDA type subunit 2D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000836.2 | Intron | NP_000827.2 | ||||
XM_011526872.1 | Intron | XP_011525174.1 |
KDELR1 - KDEL endoplasmic reticulum protein retention receptor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |