Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGTCCCCAGAGGTGCTGATGA[C/T]GTGAAACAGGGTGACATTGTAGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601052 | ||||||||||||||||||||
Literature Links: |
OLFM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OLFM2 - olfactomedin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304347.1 | 1499 | Missense Mutation | ATC,GTC | I,V 471 | NP_001291276.1 | |
NM_001304348.1 | 1499 | Missense Mutation | ATC,GTC | I,V 369 | NP_001291277.1 | |
NM_058164.3 | 1499 | Missense Mutation | ATC,GTC | I,V 447 | NP_477512.1 |
PIN1 - peptidylprolyl cis/trans isomerase, NIMA-interacting 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |