Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTAGGGCACCATCGGGAGGCC[A/G]ATGGGGTCATATGGAGACACACGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLEC16 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIGLEC16 - sialic acid binding Ig like lectin 16 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VRK3 - vaccinia related kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025778.1 | 1891 | Silent Mutation | ATC,ATT | I,I 418 | NP_001020949.1 | |
NM_001308420.1 | 1891 | Intron | NP_001295349.1 | |||
NM_016440.3 | 1891 | Silent Mutation | ATC,ATT | I,I 468 | NP_057524.3 | |
XM_005258971.3 | 1891 | Silent Mutation | ATC,ATT | I,I 468 | XP_005259028.1 | |
XM_005258972.3 | 1891 | Silent Mutation | ATC,ATT | I,I 418 | XP_005259029.1 | |
XM_006723237.3 | 1891 | Silent Mutation | ATC,ATT | I,I 468 | XP_006723300.1 | |
XM_006723238.3 | 1891 | Silent Mutation | ATC,ATT | I,I 418 | XP_006723301.1 | |
XM_011527023.2 | 1891 | Intron | XP_011525325.1 |