Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGCCAACCACTGGGACCTGGAG[C/T]TGCTCACACCAGGGAACCTGGAACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616759 MIM: 616633 MIM: 604429 | ||||||||||||||||||||
Literature Links: |
NOSIP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOSIP - nitric oxide synthase interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR12 - proline rich 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRG2 - proline rich and Gla domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000951.2 | 501 | Silent Mutation | CTG,TTG | L,L 57 | NP_000942.1 | |
NM_001316335.1 | 501 | Silent Mutation | CTG,TTG | L,L 34 | NP_001303264.1 | |
XM_006723286.2 | 501 | Silent Mutation | CTG,TTG | L,L 57 | XP_006723349.1 | |
XM_011527124.1 | 501 | Silent Mutation | CTG,TTG | L,L 34 | XP_011525426.1 |