Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGAGGGCAGGGTCCCAGTTTTG[A/T]GGAGTTGCAGAGTCCACCTATCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF773 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF773 - zinc finger protein 773 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304334.1 | 630 | Missense Mutation | AGG,TGG | R,W 135 | NP_001291263.1 | |
NM_001304335.1 | 630 | Intron | NP_001291264.1 | |||
NM_001304336.1 | 630 | UTR 3 | NP_001291265.1 | |||
NM_001304337.1 | 630 | Intron | NP_001291266.1 | |||
NM_198542.2 | 630 | Missense Mutation | AGG,TGG | R,W 136 | NP_940944.1 |