Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTCCAGGCAGCACGCTGCTCCTC[C/T]TCCTTGTGGCGCTGCTGCGCCTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606580 | ||||||||||||||||||||
Literature Links: |
OPA3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OPA3 - optic atrophy 3 (autosomal recessive, with chorea and spastic paraplegia) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017989.2 | 398 | Intron | NP_001017989.2 | |||
NM_025136.3 | 398 | Silent Mutation | GAA,GAG | E,E 112 | NP_079412.1 | |
XM_006723403.3 | 398 | Silent Mutation | GAA,GAG | E,E 59 | XP_006723466.1 |