Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGCTGTGAATTCTCCAGTGTA[C/T]ATTAAGGTGGGAGCTTTGGCTAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF671 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF671 - zinc finger protein 671 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321375.1 | 1916 | Missense Mutation | ATA,GTA | I,V 344 | NP_001308304.1 | |
NM_001321376.1 | 1916 | Missense Mutation | ATA,GTA | I,V 365 | NP_001308305.1 | |
NM_024833.2 | 1916 | Missense Mutation | ATA,GTA | I,V 442 | NP_079109.2 | |
XM_017027314.1 | 1916 | Missense Mutation | ATA,GTA | I,V 365 | XP_016882803.1 |