Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGGATGAGTTGAGCAAGTTCA[A/G]GTATCTGATCACGACCTTCTCCCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609364 MIM: 609661 | ||||||||||||||||||||
Literature Links: |
NLRP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NLRP2 - NLR family pyrin domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174081.1 | 222 | Missense Mutation | AAG,AGG | K,R 27 | NP_001167552.1 | |
NM_001174082.1 | 222 | Missense Mutation | AAG,AGG | K,R 27 | NP_001167553.1 | |
NM_001174083.1 | 222 | Missense Mutation | AAG,AGG | K,R 27 | NP_001167554.1 | |
NM_017852.3 | 222 | Missense Mutation | AAG,AGG | K,R 27 | NP_060322.1 |
NLRP7 - NLR family pyrin domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |