Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTGCGAGGGCTGCGGGAACGCC[A/G]GGAGGGAGCTTCCCGGCCCCGTCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612256 MIM: 609553 | ||||||||||||||||||||
Literature Links: |
MAST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAST1 - microtubule associated serine/threonine kinase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTBDN - retbindin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080997.2 | 763 | Missense Mutation | CGG,TGG | R,W 204 | NP_001074466.1 | |
NM_001270440.1 | 763 | UTR 3 | NP_001257369.1 | |||
NM_001270441.1 | 763 | Missense Mutation | CGG,TGG | R,W 214 | NP_001257370.1 | |
NM_001270442.1 | 763 | Missense Mutation | CGG,TGG | R,W 210 | NP_001257371.1 | |
NM_001270443.1 | 763 | Missense Mutation | CGG,TGG | R,W 204 | NP_001257372.1 | |
NM_001270444.1 | 763 | Missense Mutation | CGG,TGG | R,W 204 | NP_001257373.1 | |
NM_001270445.1 | 763 | Missense Mutation | CGG,TGG | R,W 198 | NP_001257374.1 | |
NM_031429.2 | 763 | Missense Mutation | CGG,TGG | R,W 236 | NP_113617.1 | |
XM_005260089.1 | 763 | Missense Mutation | CGG,TGG | R,W 261 | XP_005260146.1 | |
XM_005260090.3 | 763 | Missense Mutation | CGG,TGG | R,W 239 | XP_005260147.1 | |
XM_011528343.1 | 763 | Missense Mutation | CGG,TGG | R,W 256 | XP_011526645.1 | |
XM_011528344.2 | 763 | Missense Mutation | CGG,TGG | R,W 239 | XP_011526646.1 | |
XM_011528345.1 | 763 | Missense Mutation | CGG,TGG | R,W 229 | XP_011526647.1 | |
XM_017027343.1 | 763 | Missense Mutation | CGG,TGG | R,W 246 | XP_016882832.1 | |
XM_017027344.1 | 763 | Missense Mutation | CGG,TGG | R,W 230 | XP_016882833.1 |