Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGGCCGAGTACCTCACCGTGG[A/C]CATGCTCATGATTGGGGTCTTTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612891 MIM: 603014 | ||||||||||||||||||||
Literature Links: |
LRRC8E PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LRRC8E - leucine rich repeat containing 8 family member E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001268284.2 | 345 | Missense Mutation | GAC,GCC | D,A 34 | NP_001255213.1 | |
NM_001268285.2 | 345 | Intron | NP_001255214.1 | |||
NM_025061.5 | 345 | Missense Mutation | GAC,GCC | D,A 34 | NP_079337.2 | |
XM_011528319.2 | 345 | Missense Mutation | GAC,GCC | D,A 34 | XP_011526621.1 |
MAP2K7 - mitogen-activated protein kinase kinase 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |