Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAATAAAATAAAATAAAATAAAA[C/T]AAAAATAAAATGAGTAACTCCTAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600283 | ||||||||||||||||||||
Literature Links: |
GRIK5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRIK5 - glutamate ionotropic receptor kainate type subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF574 - zinc finger protein 574 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022752.5 | Intron | NP_073589.4 | ||||
XM_005259159.2 | Intron | XP_005259216.1 | ||||
XM_011527228.2 | Intron | XP_011525530.1 | ||||
XM_011527229.2 | Intron | XP_011525531.1 | ||||
XM_011527230.2 | Intron | XP_011525532.1 | ||||
XM_017027149.1 | Intron | XP_016882638.1 |