Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCAAGTAGTCAGGCGTCTCCGA[C/T]GTGGACGTGGGAACACTGGAAACGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600284 MIM: 611670 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ELL - elongation factor for RNA polymerase II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006532.3 | 757 | Silent Mutation | ACA,ACG | T,T 503 | NP_006523.1 | |
XM_011528330.2 | 757 | Intron | XP_011526632.2 | |||
XM_017027335.1 | 757 | Silent Mutation | ACA,ACG | T,T 370 | XP_016882824.1 | |
XM_017027336.1 | 757 | Silent Mutation | ACA,ACG | T,T 252 | XP_016882825.1 | |
XM_017027337.1 | 757 | Silent Mutation | ACA,ACG | T,T 219 | XP_016882826.1 |
ISYNA1 - inositol-3-phosphate synthase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |