Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCTTGTGCACAGAAAGATAGGA[A/G]TTACTTGTGAAGGATTTCCCACAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF558 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF558 - zinc finger protein 558 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304350.1 | 2279 | Silent Mutation | AAC,AAT | N,N 317 | NP_001291279.1 | |
NM_144693.2 | 2279 | Silent Mutation | AAC,AAT | N,N 388 | NP_653294.1 | |
XM_011527703.2 | 2279 | Silent Mutation | AAC,AAT | N,N 317 | XP_011526005.1 | |
XM_017026370.1 | 2279 | Silent Mutation | AAC,AAT | N,N 317 | XP_016881859.1 | |
XM_017026371.1 | 2279 | Silent Mutation | AAC,AAT | N,N 317 | XP_016881860.1 | |
XM_017026372.1 | 2279 | Silent Mutation | AAC,AAT | N,N 317 | XP_016881861.1 |