Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGTTGGAAAACTATTGCCACTT[C/T]GTATCTGTGGGTAAGAAACACTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609516 | ||||||||||||||||||||
Literature Links: |
ZNF382 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF382 - zinc finger protein 382 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256838.1 | 231 | Silent Mutation | TTC,TTT | F,F 42 | NP_001243767.1 | |
NM_032825.4 | 231 | Silent Mutation | TTC,TTT | F,F 43 | NP_116214.2 |
ZNF529 - zinc finger protein 529 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |