Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCTCTGCCCCAGACACTCACAC[A/G]GACCCTATAGAATCCTATATCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD33 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD33 - CD33 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001082618.1 | 912 | Intron | NP_001076087.1 | |||
NM_001177608.1 | 912 | Intron | NP_001171079.1 | |||
NM_001772.3 | 912 | Intron | NP_001763.3 | |||
XM_011527531.2 | 912 | UTR 5 | XP_011525833.1 | |||
XM_011527532.2 | 912 | UTR 5 | XP_011525834.1 | |||
XM_017027508.1 | 912 | UTR 5 | XP_016882997.1 | |||
XM_017027509.1 | 912 | UTR 5 | XP_016882998.1 | |||
XM_017027510.1 | 912 | UTR 5 | XP_016882999.1 |