Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGAGACCAGCTGCATCTCAACC[A/G]TAATAACATCAGAGAACCCAGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191041 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR7975 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR7975 - microRNA 7975 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNNT1 - troponin T1, slow skeletal type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126132.2 | 1166 | UTR 3 | NP_001119604.1 | |||
NM_001126133.2 | 1166 | UTR 3 | NP_001119605.1 | |||
NM_001291774.1 | 1166 | UTR 3 | NP_001278703.1 | |||
NM_003283.5 | 1166 | UTR 3 | NP_003274.3 | |||
XM_011527246.2 | 1166 | UTR 3 | XP_011525548.1 | |||
XM_017027186.1 | 1166 | UTR 3 | XP_016882675.1 | |||
XM_017027187.1 | 1166 | UTR 3 | XP_016882676.1 |