Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCTCCAAGCGGGTTGGGGTGCC[C/T]GGCATCTCAAGGTCACGTTGACCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603832 MIM: 606862 MIM: 609519 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NDUFA3 - NADH:ubiquinone oxidoreductase subunit A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OSCAR - osteoclast associated, immunoglobulin-like receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282349.1 | 365 | Missense Mutation | CAG,CGG | Q,R 54 | NP_001269278.1 | |
NM_001282350.1 | 365 | Missense Mutation | CAG,CGG | Q,R 43 | NP_001269279.1 | |
NM_130771.4 | 365 | Missense Mutation | CAG,CGG | Q,R 58 | NP_570127.3 | |
NM_133168.4 | 365 | Missense Mutation | CAG,CGG | Q,R 43 | NP_573398.2 | |
NM_133169.4 | 365 | Missense Mutation | CAG,CGG | Q,R 54 | NP_573399.2 | |
NM_206818.2 | 365 | Missense Mutation | CAG,CGG | Q,R 58 | NP_996554.2 |
TFPT - TCF3 fusion partner | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |