Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATACAAGGACACCAGTTTCTCCA[A/C]TTGCTCGCCTGGCAGGTTTTAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610823 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC25A42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC25A42 - solute carrier family 25 member 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321544.1 | Intron | NP_001308473.1 | ||||
NM_178526.4 | Intron | NP_848621.2 | ||||
XM_005259861.3 | Intron | XP_005259918.1 | ||||
XM_011527937.2 | Intron | XP_011526239.1 | ||||
XM_017026670.1 | Intron | XP_016882159.1 |