Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGGCCCATATATAATTCTTTGAC[C/T]GTCTACTACATATGATGTAATGTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC284344 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC284344 - uncharacterized LOC284344 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSG4 - pregnancy specific beta-1-glycoprotein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276495.1 | 494 | Missense Mutation | CGT,TGT | R,C 86 | NP_001263424.1 | |
NM_001316339.1 | 494 | Intron | NP_001303268.1 | |||
NM_002780.4 | 494 | Missense Mutation | CGT,TGT | R,C 86 | NP_002771.2 | |
NM_213633.2 | 494 | Missense Mutation | CGT,TGT | R,C 86 | NP_998798.1 | |
XM_017026995.1 | 494 | Intron | XP_016882484.1 | |||
XM_017026996.1 | 494 | Missense Mutation | CGT,TGT | R,C 102 | XP_016882485.1 | |
XM_017026997.1 | 494 | Missense Mutation | CGT,TGT | R,C 86 | XP_016882486.1 | |
XM_017026998.1 | 494 | Intron | XP_016882487.1 | |||
XM_017026999.1 | 494 | Intron | XP_016882488.1 | |||
XM_017027000.1 | 494 | Missense Mutation | CGT,TGT | R,C 102 | XP_016882489.1 | |
XM_017027001.1 | 494 | Intron | XP_016882490.1 | |||
XM_017027002.1 | 494 | Intron | XP_016882491.1 |