Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAGACATACAACTCACTGGCTGC[C/T]TGGCTGACGGATGCCCGCACCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601340 MIM: 612945 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIA - melanoma inhibitory activity | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIA-RAB4B - MIA-RAB4B readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAB4B - RAB4B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016154.4 | 460 | Silent Mutation | GCC,GCT | A,A 101 | NP_057238.3 |
RAB4B-EGLN2 - RAB4B-EGLN2 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |