Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTAGAGAAAACCATCCAAAAGTAC[C/G]GCACCAACCCCGAGGAGACCGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 151523 MIM: 607972 MIM: 601729 | ||||||||||||||||||||
Literature Links: |
CD37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD37 - CD37 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040031.1 | 510 | Missense Mutation | CGC,GGC | R,G 63 | NP_001035120.1 | |
NM_001774.2 | 510 | Missense Mutation | CGC,GGC | R,G 131 | NP_001765.1 | |
XM_005259435.3 | 510 | Missense Mutation | CGC,GGC | R,G 131 | XP_005259492.1 | |
XM_005259436.3 | 510 | Missense Mutation | CGC,GGC | R,G 63 | XP_005259493.1 | |
XM_011527542.2 | 510 | Missense Mutation | CGC,GGC | R,G 63 | XP_011525844.1 | |
XM_011527543.2 | 510 | Missense Mutation | CGC,GGC | R,G 63 | XP_011525845.1 | |
XM_011527544.2 | 510 | Missense Mutation | CGC,GGC | R,G 63 | XP_011525846.1 | |
XM_017027513.1 | 510 | Missense Mutation | CGC,GGC | R,G 63 | XP_016883002.1 |
SLC6A16 - solute carrier family 6 member 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEAD2 - TEA domain transcription factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |