Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACAGCATCTGTGGATTTAGGTAT[A/G]CTTAGGGGGTCTGGAACCAATCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF442 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF442 - zinc finger protein 442 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030824.2 | 2089 | UTR 3 | NP_110451.1 | |||
XM_006722908.3 | 2089 | UTR 3 | XP_006722971.1 | |||
XM_017027316.1 | 2089 | UTR 3 | XP_016882805.1 | |||
XM_017027317.1 | 2089 | UTR 3 | XP_016882806.1 | |||
XM_017027318.1 | 2089 | UTR 3 | XP_016882807.1 |
ZNF563 - zinc finger protein 563 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |