Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTTACCTCATGCAAAGAACCTTC[A/G]GTTAAATTAGGGACACTTCTATCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ANKRD27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD27 - ankyrin repeat domain 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032139.2 | 3057 | Silent Mutation | ACC,ACT | T,T 967 | NP_115515.2 | |
XM_006723412.2 | 3057 | Silent Mutation | ACC,ACT | T,T 967 | XP_006723475.1 | |
XM_017027349.1 | 3057 | Silent Mutation | ACC,ACT | T,T 771 | XP_016882838.1 |