Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTACGATGAGAACTGGTTCTACA[C/T]GCGAGCTGGTGAGGAACTTAGGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614806 MIM: 603474 | ||||||||||||||||||||
Literature Links: |
DMRTC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DMRTC2 - DMRT like family C2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6797 - microRNA 6797 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS19 - ribosomal protein S19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001022.3 | 536 | Missense Mutation | ACG,ATG | T,M 55 | NP_001013.1 | |
NM_001321483.1 | 536 | Intron | NP_001308412.1 | |||
NM_001321484.1 | 536 | Intron | NP_001308413.1 | |||
NM_001321485.1 | 536 | Missense Mutation | ACG,ATG | T,M 55 | NP_001308414.1 | |
XM_017027113.1 | 536 | Missense Mutation | ACG,ATG | T,M 55 | XP_016882602.1 |