Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGTAGACGCAGCTGTAGTTGGC[A/G]GAGTCAGCCACGGAAATGTTGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138670 MIM: 165260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
A1BG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
A1BG - alpha-1-B glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_130786.3 | 1169 | Silent Mutation | TCC,TCT | S,S 369 | NP_570602.2 |
A1BG-AS1 - A1BG antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100419840 - zinc finger protein 446 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF497 - zinc finger protein 497 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN22 - zinc finger and SCAN domain containing 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |