Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCGGGAAGTTCTGCTCCACGAG[A/G]AGCTGGGCCAGGGCCATGCAGCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601211 | ||||||||||||||||||||
Literature Links: |
ADGRE5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADGRE5 - adhesion G protein-coupled receptor E5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025160.2 | 1033 | Intron | NP_001020331.1 | |||
NM_001784.4 | 1033 | Intron | NP_001775.2 | |||
NM_078481.3 | 1033 | Intron | NP_510966.1 | |||
XM_011528451.2 | 1033 | Intron | XP_011526753.1 | |||
XM_017027547.1 | 1033 | Intron | XP_016883036.1 |
DDX39A - DEAD-box helicase 39A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005804.3 | 1033 | Silent Mutation | CTC,CTT | L,L 305 | NP_005795.2 | |
XM_006722606.3 | 1033 | Silent Mutation | CTC,CTT | L,L 100 | XP_006722669.1 | |
XM_011527620.1 | 1033 | Silent Mutation | CTC,CTT | L,L 305 | XP_011525922.1 | |
XM_011527621.2 | 1033 | Silent Mutation | CTC,CTT | L,L 100 | XP_011525923.1 |