Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAATTCTCTGATGCCTTGCAAGTT[C/T]TGAAGTTTGACTAAAGGCTTTGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF347 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF347 - zinc finger protein 347 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172674.1 | 2765 | Missense Mutation | AAA,GAA | K,E 780 | NP_001166145.1 | |
NM_001172675.1 | 2765 | Missense Mutation | AAA,GAA | K,E 780 | NP_001166146.1 | |
NM_032584.2 | 2765 | Missense Mutation | AAA,GAA | K,E 779 | NP_115973.2 | |
XM_005259335.4 | 2765 | Missense Mutation | AAA,GAA | K,E 779 | XP_005259392.1 | |
XM_005259336.4 | 2765 | Missense Mutation | AAA,GAA | K,E 721 | XP_005259393.1 | |
XM_017027384.1 | 2765 | Missense Mutation | AAA,GAA | K,E 779 | XP_016882873.1 | |
XM_017027385.1 | 2765 | Missense Mutation | AAA,GAA | K,E 721 | XP_016882874.1 | |
XM_017027386.1 | 2765 | Missense Mutation | AAA,GAA | K,E 721 | XP_016882875.1 |
ZNF415 - zinc finger protein 415 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |