Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTCACGTCCCTGTCTGTCCTGTC[A/G]AGAGGGGGAAGCATCCCAGACCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607287 MIM: 608239 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSM7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LSM7 - LSM7 homolog, U6 small nuclear RNA and mRNA degradation associated | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPPL2B - signal peptide peptidase like 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077238.1 | Intron | NP_001070706.1 | ||||
NM_152988.2 | Intron | NP_694533.1 | ||||
XM_011528138.2 | Intron | XP_011526440.1 | ||||
XM_011528139.1 | Intron | XP_011526441.1 | ||||
XM_011528140.1 | Intron | XP_011526442.1 | ||||
XM_017027016.1 | Intron | XP_016882505.1 | ||||
XM_017027017.1 | Intron | XP_016882506.1 | ||||
XM_017027018.1 | Intron | XP_016882507.1 | ||||
XM_017027019.1 | Intron | XP_016882508.1 |