Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCCTTGCAGCCTCACAGAATGAG[A/G]TTCCTGCGGCACCCCTGGAAGAACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MUM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MUM1 - melanoma associated antigen (mutated) 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032853.3 | 332 | Missense Mutation | ATT,GTT | I,V 78 | NP_116242.2 | |
XM_011528380.2 | 332 | Missense Mutation | ATT,GTT | I,V 78 | XP_011526682.1 | |
XM_011528382.2 | 332 | Missense Mutation | ATT,GTT | I,V 78 | XP_011526684.1 | |
XM_011528383.1 | 332 | Missense Mutation | ATT,GTT | I,V 78 | XP_011526685.1 | |
XM_011528384.2 | 332 | Missense Mutation | ATT,GTT | I,V 78 | XP_011526686.1 | |
XM_011528385.2 | 332 | Missense Mutation | ATT,GTT | I,V 9 | XP_011526687.1 | |
XM_017027401.1 | 332 | Missense Mutation | ATT,GTT | I,V 78 | XP_016882890.1 |