Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCAAGACATGGGTAGCGTCCAA[C/T]GAGATCAAGGACAAGAGGCAGCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNOT3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNOT3 - CCR4-NOT transcription complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014516.3 | 565 | Silent Mutation | AAC,AAT | N,N 69 | NP_055331.1 | |
XM_005278279.1 | 565 | Silent Mutation | AAC,AAT | N,N 69 | XP_005278336.1 | |
XM_005278280.2 | 565 | Silent Mutation | AAC,AAT | N,N 69 | XP_005278337.1 | |
XM_005278281.1 | 565 | Silent Mutation | AAC,AAT | N,N 69 | XP_005278338.1 | |
XM_005278282.2 | 565 | Silent Mutation | AAC,AAT | N,N 69 | XP_005278339.1 | |
XM_011526992.1 | 565 | Silent Mutation | AAC,AAT | N,N 69 | XP_011525294.1 | |
XM_011526993.2 | 565 | Silent Mutation | AAC,AAT | N,N 69 | XP_011525295.1 |