Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAGTAGAGCAGAAGAAGAAGCG[A/G]ACCTTCCGCAAGTTCACCTACCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612034 MIM: 607430 MIM: 180535 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APC2 - APC2, WNT signaling pathway regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DAZAP1 - DAZ associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPS15 - ribosomal protein S15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018.4 | Intron | NP_001009.1 | ||||
NM_001308226.1 | Intron | NP_001295155.1 |