Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGCCTTACTTCCTGGAGGAGCC[C/T]GAAGACAGGACTGTGGCCGCCAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 109135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AXL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AXL - AXL receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278599.1 | 629 | Intron | NP_001265528.1 | |||
NM_001699.5 | 629 | Silent Mutation | CCC,CCT | P,P 145 | NP_001690.2 | |
NM_021913.4 | 629 | Silent Mutation | CCC,CCT | P,P 145 | NP_068713.2 |