Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACACCCTCTTACCTGCGCCCTCC[A/G]GATCGCTTCCTGCAGCTCCTCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 182100 MIM: 610349 MIM: 609623 | ||||||||||||||||||||
Literature Links: |
FUT2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FUT2 - fucosyltransferase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAMSTR - MEF2 activating motif and SAP domain containing transcriptional regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130915.1 | 1728 | Missense Mutation | CGG,TGG | R,W 300 | NP_001124387.1 | |
NM_001297753.1 | 1728 | Missense Mutation | CGG,TGG | R,W 132 | NP_001284682.1 | |
NM_182574.2 | 1728 | Missense Mutation | CGG,TGG | R,W 197 | NP_872380.1 | |
XM_011526807.2 | 1728 | Missense Mutation | CGG,TGG | R,W 197 | XP_011525109.1 | |
XM_011526808.2 | 1728 | Missense Mutation | CGG,TGG | R,W 363 | XP_011525110.1 | |
XM_011526809.2 | 1728 | Missense Mutation | CGG,TGG | R,W 132 | XP_011525111.1 | |
XM_017026640.1 | 1728 | Missense Mutation | CGG,TGG | R,W 381 | XP_016882129.1 | |
XM_017026641.1 | 1728 | Missense Mutation | CGG,TGG | R,W 368 | XP_016882130.1 | |
XM_017026642.1 | 1728 | Missense Mutation | CGG,TGG | R,W 197 | XP_016882131.1 | |
XM_017026643.1 | 1728 | Missense Mutation | CGG,TGG | R,W 328 | XP_016882132.1 | |
XM_017026644.1 | 1728 | Missense Mutation | CGG,TGG | R,W 318 | XP_016882133.1 | |
XM_017026645.1 | 1728 | Missense Mutation | CGG,TGG | R,W 132 | XP_016882134.1 |
RASIP1 - Ras interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |