Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCCACTGACAGCCGCCAGCCC[A/G]GGCGAGCTGCCCACCGAGGGTGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605414 MIM: 601155 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCA7 - ATP binding cassette subfamily A member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ARHGAP45 - Rho GTPase activating protein 45 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258328.2 | 387 | Silent Mutation | CCA,CCG | P,P 116 | NP_001245257.1 | |
NM_001282334.1 | 387 | Intron | NP_001269263.1 | |||
NM_001282335.1 | 387 | Intron | NP_001269264.1 | |||
NM_001321232.1 | 387 | Silent Mutation | CCA,CCG | P,P 104 | NP_001308161.1 | |
NM_012292.4 | 387 | Silent Mutation | CCA,CCG | P,P 100 | NP_036424.2 | |
XM_006722713.1 | 387 | Silent Mutation | CCA,CCG | P,P 100 | XP_006722776.1 | |
XM_011527858.1 | 387 | Silent Mutation | CCA,CCG | P,P 104 | XP_011526160.1 | |
XM_011527860.2 | 387 | Intron | XP_011526162.1 |